Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012203.1 | NP_001012203.1 | 1 - 2 | 88 | 101 | |
GenBank mRNA | BC079035.1 | - | 1 - 2 | 88 | 7114311 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |