Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017209.1 | NP_058905.1 | 2 - 3 | 299 | 76 | |
XM_006239009.2 | XP_006239071.1 | 3 - 4 | 418 | 76 | ||
GenBank mRNA | AB015724.1 | - | 2 - 3 | 299 | 76 | |
FQ210707.1 | - | 2 - 3 | 295 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |