Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249121.2 | XP_006249183.1 | 1 - 2 | 371 | 68 | |
XM_006249123.2 | XP_006249185.1 | 1 - 2 | 351 | 68 | ||
GenBank mRNA | FQ213833.1 | - | 1 - 2 | 131 | 68 | |
FQ222246.1 | - | 1 - 2 | 117 | 68 | ||
FQ224250.1 | - | 1 - 2 | 128 | 68 | ||
FQ232488.1 | - | 1 - 2 | 117 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |