Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017077.2 | NP_058773.1 | 1 - 2 | 145 | 64 | |
GenBank mRNA | L09648.1 | - | 1 - 2 | 146 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |