Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107758.1 | NP_001101228.1 | 1 - 2 | 249 | 66 | |
XM_006234747.2 | XP_006234809.1 | 1 - 2 | 236 | 66 | ||
XM_006234749.2 | XP_006234811.1 | 1 - 2 | 236 | 66 | ||
XM_006234752.2 | XP_006234814.1 | 1 - 2 | 236 | 66 | ||
XM_006234754.2 | XP_006234816.1 | 1 - 2 | 236 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |