Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108676.1 | NP_001102146.1 | 1 - 2 | 184 | 75 | |
XM_006238804.2 | XP_006238866.1 | 1 - 2 | 765 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |