Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khdrbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130405.1 | NP_569089.1 | 3 - 4 | 705 | 78 | |
GenBank mRNA | AF305619.1 | - | 3 - 4 | 705 | 78 | |
AF393783.1 | - | 3 - 4 | 622 | 78 | ||
BC061987.1 | - | 3 - 4 | 729 | 5599491 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |