Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmfg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181091.2 | NP_851605.1 | 1 - 2 | 105 | 65 | |
XM_006228606.2 | XP_006228668.1 | 2 - 3 | 403 | 65 | ||
XM_006228607.2 | XP_006228669.1 | 2 - 3 | 403 | 65 | ||
XM_008759100.1 | XP_008757322.1 | 2 - 3 | 218 | 65 | ||
GenBank mRNA | AB007364.1 | - | 1 - 2 | 105 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |