Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012506.1 | NP_036638.1 | 15 - 16 | 2236 | 74 | |
XM_008758888.1 | XP_008757110.1 | 15 - 16 | 2150 | 74 | ||
GenBank mRNA | FQ212331.1 | - | 14 - 15 | 2073 | 74 | |
M14513.1 | - | 15 - 16 | 2236 | 74 | ||
M28648.1 | - | 14 - 15 | 2090 | 74 | ||
X05883.1 | - | 15 - 16 | 2233 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |