Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145677.1 | NP_663710.1 | 7 - 8 | 1008 | 56 | |
XM_006233931.2 | XP_006233993.1 | 8 - 9 | 1165 | 56 | ||
XM_006233932.2 | XP_006233994.1 | 7 - 8 | 1129 | 56 | ||
XM_006233933.2 | XP_006233995.1 | 8 - 9 | 1817 | 56 | ||
XM_006233934.2 | XP_006233996.1 | 7 - 8 | 1781 | 56 | ||
XM_006233935.2 | XP_006233997.1 | 8 - 9 | 2381 | 56 | ||
XM_006233937.2 | XP_006233999.1 | 8 - 9 | 1401 | 56 | ||
GenBank mRNA | AY043169.1 | - | 7 - 8 | 1008 | 56 | |
FQ211394.1 | - | 7 - 8 | 1088 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |