Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012510.2 | NP_036642.2 | 3 - 4 | 534 | 55 | |
XM_008771385.1 | XP_008769607.1 | 3 - 4 | 359 | 55 | ||
GenBank mRNA | M35535.1 | - | 3 - 4 | 534 | 55 | |
M55655.1 | - | 3 - 4 | 370 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |