Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012510.2 | NP_036642.2 | 4 - 5 | 736 | 92 | |
XM_008771385.1 | XP_008769607.1 | 4 - 5 | 561 | 92 | ||
GenBank mRNA | M35535.1 | - | 4 - 5 | 736 | 92 | |
M55655.1 | - | 4 - 5 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |