Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012526.2 | NP_036658.2 | 1 - 2 | 311 | 140 | |
GenBank mRNA | AF019974.1 | - | 1 - 2 | 111 | 140 | |
BC128743.1 | - | 1 - 2 | 130 | 8370781 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |