Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012692.1 | NP_036824.1 | 3 - 4 | 358 | 63 | |
XM_008759105.1 | XP_008757327.1 | 3 - 4 | 386 | 63 | ||
XM_008759106.1 | XP_008757328.1 | 3 - 4 | 386 | 63 | ||
GenBank mRNA | BC081848.1 | - | 3 - 4 | 348 | 7135880 | 63 |
J02669.1 | - | 3 - 4 | 358 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection P450 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |