Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012826.2 | NP_036958.2 | 3 - 4 | 618 | 79 | |
XM_006249003.2 | XP_006249065.1 | 5 - 6 | 799 | 79 | ||
XM_006249004.2 | XP_006249066.1 | 5 - 6 | 783 | 79 | ||
GenBank mRNA | BC105822.1 | - | 3 - 4 | 618 | 7371444 | 79 |
D21058.1 | - | 3 - 4 | 617 | 79 | ||
FQ210978.1 | - | 637 | 79 | |||
X75309.1 | - | 3 - 4 | 599 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |