Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012909.2 | NP_037041.2 | 1 - 2 | 448 | 111 | |
GenBank mRNA | AF110021.1 | - | 1 - 2 | 351 | 111 | |
BC128705.1 | - | 1 - 2 | 434 | 7109642 | 111 | |
D13906.1 | - | 1 - 2 | 448 | 111 | ||
L28112.1 | - | 1 - 2 | 487 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |