Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013064.1 | NP_037196.1 | 8 - 9 | 1654 | 68 | |
GenBank mRNA | AF041244.1 | - | 8 - 9 | 1654 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |