Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013092.1 | NP_037224.1 | 1 - 2 | 90 | 83 | |
GenBank mRNA | D38495.1 | - | 1 - 2 | 90 | 83 | |
U67908.1 | - | 1 - 2 | 55 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protease Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons |