Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013097.2 | NP_037229.1 | 3 - 4 | 514 | 70 | |
XM_006245829.2 | XP_006245891.1 | 4 - 5 | 1194 | 70 | ||
GenBank mRNA | AF397150.1 | - | 3 - 4 | 514 | 70 | |
AF397151.1 | - | 4 - 5 | 385 | 70 | ||
BC089764.1 | - | 3 - 4 | 514 | 7130586 | 70 | |
X56060.1 | - | 3 - 4 | 367 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |