Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcyap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016989.2 | NP_058685.1 | 4 - 5 | 632 | 94 | |
XM_006245672.2 | XP_006245734.1 | 5 - 6 | 1328 | 94 | ||
XM_006245673.2 | XP_006245735.1 | 5 - 6 | 1704 | 94 | ||
XM_006245674.2 | XP_006245736.1 | 5 - 6 | 1657 | 94 | ||
XM_008767418.1 | XP_008765640.1 | 5 - 6 | 1281 | 94 | ||
XM_008767419.1 | XP_008765641.1 | 3 - 4 | 678 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |