Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017030.2 | NP_058726.2 | 10 - 11 | 1101 | 125 | |
GenBank mRNA | BC078855.1 | - | 10 - 11 | 1101 | 7108495 | 125 |
M14634.1 | - | 10 - 11 | 1102 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |