Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capza3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017164.2 | NP_058860.2 | 1 | 370 | 65 | |
GenBank mRNA | BC078870.1 | - | 1 | 370 | 7133258 | 65 |
Y12538.1 | - | 1 | 350 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |