Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017182.2 | NP_058878.1 | 1 - 2 | 149 | 80 | |
XM_006253573.2 | XP_006253635.1 | 1 - 2 | 184 | 80 | ||
XM_006253575.2 | XP_006253637.1 | 1 - 2 | 184 | 80 | ||
XM_006253576.2 | XP_006253638.1 | 1 - 2 | 184 | 80 | ||
XM_006253577.2 | XP_006253639.1 | 1 - 2 | 184 | 80 | ||
GenBank mRNA | BC089093.1 | - | 1 - 2 | 149 | 7126329 | 80 |
U79139.1 | - | 1 - 2 | 94 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |