Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017311.1 | NP_059007.1 | 2 - 3 | 98 | 94 | |
GenBank mRNA | D13123.1 | - | 2 - 3 | 98 | 94 | |
FQ209790.1 | - | 2 - 3 | 116 | 94 | ||
FQ216886.1 | - | 2 - 3 | 116 | 94 | ||
FQ217997.1 | - | 2 - 3 | 112 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Rattus norvegicus Inventoried Probe spans exons |