Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017362.4 | NP_059058.1 | 892 | 58 | ||
GenBank mRNA | M22926.1 | - | 892 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Ion Channel Rattus norvegicus Inventoried Probe spans exons |