Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019158.2 | NP_062031.1 | 2 - 3 | 390 | 87 | |
XM_006230187.2 | XP_006230249.1 | 2 - 3 | 437 | 87 | ||
GenBank mRNA | AB005547.1 | - | 2 - 3 | 389 | 87 | |
AF007775.1 | - | 2 - 3 | 369 | 87 | ||
BC081812.1 | - | 2 - 3 | 390 | 7114305 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |