Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019182.2 | NP_062055.1 | 3 - 4 | 411 | 62 | |
XM_008770271.1 | XP_008768493.1 | 3 - 4 | 443 | 62 | ||
XM_008770272.1 | XP_008768494.1 | 3 - 4 | 398 | 62 | ||
XM_008770273.1 | XP_008768495.1 | 3 - 4 | 531 | 62 | ||
GenBank mRNA | AF022081.1 | - | 3 - 4 | 329 | 62 | |
AY050655.1 | - | 3 - 4 | 423 | 62 | ||
BC062024.1 | - | 3 - 4 | 411 | 5623495 | 62 | |
FQ211855.1 | - | 2 - 3 | 293 | 62 | ||
FQ223606.1 | - | 3 - 4 | 427 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons |