Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127339.1 | NP_001120811.1 | 3 - 4 | 471 | 58 | |
NM_001127341.1 | NP_001120813.1 | 3 - 4 | 471 | 58 | ||
NM_019221.3 | NP_062094.1 | 3 - 4 | 471 | 58 | ||
XM_006248500.2 | XP_006248562.1 | 3 - 4 | 552 | 58 | ||
XM_008768791.1 | XP_008767013.1 | 4 - 5 | 401 | 58 | ||
GenBank mRNA | AJ277446.1 | - | 2 - 3 | 273 | 58 | |
AJ277448.1 | - | 2 - 3 | 273 | 58 | ||
AJ277449.1 | - | 3 - 4 | 324 | 58 | ||
AJ277451.1 | - | 3 - 4 | 324 | 58 | ||
AJ277452.1 | - | 2 - 3 | 273 | 58 | ||
Y10258.2 | - | 3 - 4 | 471 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Rattus norvegicus Inventoried Probe spans exons |