Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019298.1 | NP_062171.1 | 4 - 5 | 355 | 67 | |
GenBank mRNA | X74835.1 | - | 4 - 5 | 355 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |