Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019381.2 | NP_062254.2 | 3 - 4 | 242 | 58 | |
XM_006257323.2 | XP_006257385.1 | 3 - 4 | 384 | 58 | ||
XM_008765670.1 | XP_008763892.1 | 4 - 5 | 342 | 58 | ||
GenBank mRNA | AY321320.1 | - | 5 - 6 | 380 | 58 | |
AY325130.1 | - | 5 - 6 | 380 | 58 | ||
AY325243.1 | - | 5 - 6 | 380 | 58 | ||
BC058478.1 | - | 3 - 4 | 248 | 6888036 | 58 | |
FQ217052.1 | - | 3 - 4 | 241 | 58 | ||
FQ218664.1 | - | 3 - 4 | 244 | 58 | ||
FQ226858.1 | - | 3 - 4 | 271 | 58 | ||
FQ229652.1 | - | 3 - 4 | 243 | 58 | ||
FQ230608.1 | - | 3 - 4 | 271 | 58 | ||
FQ231817.1 | - | 3 - 4 | 249 | 58 | ||
X75855.1 | - | 3 - 4 | 274 | 58 | ||
X75856.1 | - | 2 - 3 | 158 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |