Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019385.2 | NP_062258.2 | 2 - 3 | 163 | 71 | |
GenBank mRNA | AF015264.2 | - | 2 - 3 | 140 | 71 | |
BC081809.1 | - | 2 - 3 | 157 | 71 | ||
BC161850.1 | - | 2 - 3 | 163 | 8369057 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |