Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021672.1 | NP_067704.1 | 1 - 2 | 393 | 60 | |
XM_008767704.1 | XP_008765926.1 | 2 - 3 | 310 | 60 | ||
GenBank mRNA | AF099912.1 | - | 1 - 2 | 393 | 60 | |
X81899.1 | - | 1 - 2 | 393 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |