Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prlh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022222.1 | NP_071558.1 | 1 - 2 | 90 | 98 | |
XM_006245478.2 | XP_006245540.1 | 2 - 3 | 794 | 98 | ||
GenBank mRNA | AB015418.1 | - | 1 - 2 | 90 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |