Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022228.1 | NP_071564.1 | 1 - 2 | 718 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Rat Androgen Panel Rat Estrogen Panel Inventoried Probe spans exons |