Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022537.1 | NP_071982.1 | 2 - 3 | 270 | 116 | |
XM_006253909.1 | XP_006253971.1 | 2 - 3 | 244 | 116 | ||
GenBank mRNA | AB000107.1 | - | 2 - 3 | 270 | 116 | |
BC098917.1 | - | 2 - 3 | 250 | 7445772 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |