Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022583.1 | NP_072105.1 | 1 - 2 | 358 | 133 | |
GenBank mRNA | AF159506.1 | - | 1 - 2 | 358 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |