Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023968.1 | NP_076458.1 | 1 | 853 | 65 | |
XM_006232534.2 | XP_006232596.1 | 2 | 1619 | 65 | ||
XM_008761113.1 | XP_008759335.1 | 2 | 948 | 65 | ||
GenBank mRNA | AY004257.1 | - | 1 | 853 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |