Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Birc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023987.3 | NP_076477.3 | 8 - 9 | 1806 | 64 | |
XM_006242499.2 | XP_006242561.1 | 7 - 8 | 3769 | 64 | ||
XM_006242500.2 | XP_006242562.1 | 8 - 9 | 2539 | 64 | ||
XM_006242502.2 | XP_006242564.1 | 7 - 8 | 2038 | 64 | ||
XM_006242503.2 | XP_006242565.1 | 7 - 8 | 1997 | 64 | ||
GenBank mRNA | AF183430.1 | - | 6 - 7 | 1574 | 64 | |
BC083555.1 | - | 8 - 9 | 1806 | 7112819 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |