Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234591.2 | XP_006234653.1 | 4 - 5 | 557 | 63 | |
XM_006234592.2 | XP_006234654.1 | 4 - 5 | 505 | 63 | ||
XM_006234593.2 | XP_006234655.1 | 4 - 5 | 1182 | 63 | ||
GenBank mRNA | AF315950.1 | - | 4 - 5 | 471 | 63 | |
BC157805.1 | - | 4 - 5 | 423 | 9026085 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |