Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031140.1 | NP_112402.1 | 2 - 3 | 700 | 105 | |
GenBank mRNA | BC061847.1 | - | 2 - 3 | 740 | 5621065 | 105 |
FQ234991.1 | - | 2 - 3 | 454 | 105 | ||
X62952.1 | - | 2 - 3 | 700 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |