Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011710.2 | NP_001011710.1 | 8 - 9 | 727 | 143 | |
XM_006244643.2 | XP_006244705.1 | 7 - 8 | 710 | 143 | ||
GenBank mRNA | AB009662.1 | - | 7 - 8 | 590 | 143 | |
BC078799.1 | - | 8 - 9 | 676 | 7112769 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |