Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031549.2 | NP_113737.1 | 1 - 2 | 39 | 77 | |
GenBank mRNA | BC061770.1 | - | 1 - 2 | 39 | 5621257 | 77 |
FQ212757.1 | - | 1 - 2 | 64 | 77 | ||
FQ219883.1 | - | 1 - 2 | 62 | 77 | ||
FQ220131.1 | - | 1 - 2 | 63 | 77 | ||
FQ220322.1 | - | 1 - 2 | 61 | 77 | ||
FQ220861.1 | - | 1 - 2 | 64 | 77 | ||
FQ220863.1 | - | 1 - 2 | 64 | 77 | ||
FQ223284.1 | - | 1 - 2 | 63 | 77 | ||
FQ224487.1 | - | 1 - 2 | 64 | 77 | ||
FQ229263.1 | - | 1 - 2 | 62 | 77 | ||
FQ229775.1 | - | 1 - 2 | 63 | 77 | ||
FQ229807.1 | - | 1 - 2 | 64 | 77 | ||
FQ234912.1 | - | 1 - 2 | 59 | 77 | ||
M83107.1 | - | 1 - 2 | 149 | 77 | ||
X71070.1 | - | 1 - 2 | 36 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |