Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031560.2 | NP_113748.1 | 7 - 8 | 964 | 69 | |
GenBank mRNA | AF010306.1 | - | 7 - 8 | 933 | 69 | |
BC078793.1 | - | 7 - 8 | 964 | 7134253 | 69 | |
FQ228691.1 | - | 7 - 8 | 981 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons |