Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031627.2 | NP_113815.2 | 6 - 7 | 998 | 90 | |
XM_006234533.2 | XP_006234595.1 | 7 - 8 | 1176 | 90 | ||
XM_006234534.1 | XP_006234596.1 | 7 - 8 | 1117 | 90 | ||
XM_006234535.2 | XP_006234597.1 | 7 - 8 | 1318 | 90 | ||
XM_006234537.1 | XP_006234599.1 | 6 - 7 | 788 | 90 | ||
XM_008762013.1 | XP_008760235.1 | 6 - 7 | 908 | 90 | ||
GenBank mRNA | BC088750.1 | - | 6 - 7 | 998 | 7314121 | 90 |
U11685.1 | - | 6 - 7 | 1005 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |