Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031703.1 | NP_113891.1 | 1 - 2 | 185 | 99 | |
GenBank mRNA | BC127490.1 | - | 1 - 2 | 167 | 8366493 | 99 |
D17695.1 | - | 1 - 2 | 100 | 99 | ||
L35108.1 | - | 1 - 2 | 185 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |