Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053499.1 | NP_445951.1 | 1 - 2 | 85 | 94 | |
XM_006236069.2 | XP_006236131.1 | 1 - 2 | 151 | 94 | ||
XM_006236070.2 | XP_006236132.1 | 1 - 2 | 151 | 94 | ||
GenBank mRNA | AJ292524.1 | - | 1 - 2 | 85 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |