Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bud31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053556.1 | NP_446008.1 | 2 - 3 | 275 | 84 | |
XM_006248887.1 | XP_006248949.1 | 2 - 3 | 392 | 84 | ||
XM_006248889.1 | XP_006248951.1 | 3 - 4 | 260 | 84 | ||
XM_008769003.1 | XP_008767225.1 | 3 - 4 | 489 | 84 | ||
GenBank mRNA | AF058791.1 | - | 2 - 3 | 275 | 84 | |
BC058456.1 | - | 3 - 4 | 302 | 6887068 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |