Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053787.2 | NP_446239.2 | 13 - 14 | 1931 | 67 | |
XM_006236376.2 | XP_006236438.1 | 11 - 12 | 1507 | 67 | ||
GenBank mRNA | AB032019.1 | - | 13 - 14 | 1931 | 67 | |
AF209196.1 | - | 1882 | 67 | |||
AY762624.1 | - | 12 - 13 | 1619 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Rattus norvegicus Probe spans exons |