Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053787.2 | NP_446239.2 | 1 - 2 | 273 | 80 | |
GenBank mRNA | AB032019.1 | - | 1 - 2 | 273 | 80 | |
AF209196.1 | - | 224 | 80 | |||
AY762624.1 | - | 1 - 2 | 108 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |