Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053787.2 | NP_446239.2 | 2 - 3 | 378 | 67 | |
GenBank mRNA | AB032019.1 | - | 2 - 3 | 378 | 67 | |
AF209196.1 | - | 329 | 67 | |||
AY762624.1 | - | 2 - 3 | 213 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |